Syringe priming kit
Leverandør: HAMILTON BONADUZ
PRMKIT
HAMIPRMKITEA 4500 NOK
HAMIPRMKIT
Syringe priming kit
Sprøyter Sprøyter for kromatografi
The priming kit provides the necessary tools to ensure that the syringe and needle connection is completely primed. The glass capillary tubing traps a large volume of liquid that can only be overcome by using the priming kit. The kit is compatible with 600, 700, 1700, removable needle syringes and 7000 series syringes.
Leveringsinformasjon: Kit includes one 250 µl syringe, one 6 pack of 30 gauge needles and one pack of septa.