Poly-spoon, with hollow handle
Leverandør: HAMMACHER KARL
HSN179-19
HAMMHSN179-19EA 140 NOK
HAMMHSN179-19
Poly-spoon, with hollow handle
Skjeer
Poly-spoon made of stainless-steel.
  • Double-ended: One spoon and one spatula end