Micro spoons, double-ended, Wironit steel
Leverandør: HAMMACHER KARL
HWN100-15
HAMMHWN100-15EA 510 NOK
HAMMHWN100-15
Micro spoons, double-ended, Wironit steel
Skjeer
Double-ended micro spoon made of Wironit steel (stainless steel 18/12 with 18 % chrome and 10 to 12 % nickel).