Gauze bandage, Peha®
Leverandør: HARTMANN PAUL
3040530
HART3040530EA 440 NOK
HART3040530
Gauze bandage, Peha®
Førstehjelpsutstyr Bandasjer
Gauze bandage made of non-elastic fabric (60% viscose/40% cotton).
  • With woven edges
  • Sold as rolls
Ideal for use as wound dressings or padding material. Also suitable as a carrier fabric for zinc glue dressings as well as first aid emergency bandages.