Hansker, kirurgi, Peha-micron® plus pudderfri
Leverandør: HARTMANN PAUL
9425742
HART9425742EA 3825 NOK
HART9425742
Hansker, kirurgi, Peha-micron® plus pudderfri
Hansker
These disposable gloves are made of thin and soft latex providing great tactile sensitivity, especially at the finger tips. Ideal for microsurgery.
  • Powder-free, smooth surface
  • Easy to put on due to inner polymer coating
  • Modified skin grip surface provides optimal grip
  • Suitable for double-gloving
  • Flexible and tear-resistant, anatomical fit
  • With reinforced cuffs
  • Light brown colour prevents light reflections
Colour: Light brown
EN 455
Sertifiseringer: EN 455