Rotating Sample Reducer, RPT
Leverandør: VWR Collection
HAVE205322396EA 0 NOK
HAVE205322396 HAVE205322402 HAVE205322419
Rotating Sample Reducer, RPT
Prøveinndelere
The HAVER rotating sample reducer produces small but representative sample quantities of solids and suspensions. It combines three dividing methods in one unit and achieves an accuracy of 99,9%.
  • Representative sample division at a rate of 1:8, 1:10 or 1:30
  • Integrated adjustable feeder with continuously regulated vibration intensity
  • Easier material flow caused by high rotating speed of the cone (approx. 100 U/min.)
  • Made from food grade materials - easy to clean
  • Fitted with a low maintenance drive motor with safety clutch