VWR® EML 200, testsiktrystere
Leverandør: VWR Collection
510-2043EA 36200 NOK
510-2043 510-2045 510-2046 510-2048 510-2037 510-2039 510-2049 510-2051 510-2040 510-2042 HAVE205323041 HAVE205323058 HAVE205323027 HAVE205322990 HAVE205323010
VWR® EML 200, testsiktrystere
Risteapparater Siktemaskiner
The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a cast aluminium housing and improved clamping system.
  • Digital timer: 0 to 99 minutes or constant operation
  • All shakers have a cover with inspection glass which allows samples to be viewed during sieving
EML 200 Pure units are ideal for standard analysis:
Feature two given regulated amplitudes to select from: Fine or coarse
Operation in set intervals of 10 seconds
Classic 'eco' clamping system