Test sieves
Leverandør: NEXOPART
206468758
HAVE206468758EA 6925 NOK
HAVE206468758
Test sieves
Sikter Sikter, test
Grid sieve with frame made from aluminium, with parallel cylindrical bars.
Sertifiseringer: DIN EN 933-3
Leveringsinformasjon: Supplied with serial number, inspection certificate according to DIN EN 10 204 and calibration certificate on request.