Weighing dishes, aluminium
Leverandør: HEATHROW
HS14522
HEATHS14522EA 230 NOK
HEATHS14522
Weighing dishes, aluminium
Skip Veieskip
Weighing dishes, made from light gauge aluminium are designed for moisture determination balances. Ideal for general-purpose weighing and dispensing.
  • Used as evaporating dish or dust cover
  • Lightweight, sturdy dish has no coating and is oil free
  • Slightly tapered for stacking