Spatula
Leverandør: HEATHROW
HS15909
HEATHS15909EA 105 NOK
HEATHS15909
Spatula
Spatler
This spatula is used to transfer, lift, and handle powders and granular materials.
  • Made from nickel-plated, stainless steel wire
  • Autoclavable
Mirror finish with flat and spoon ends.