Racks for cryogenic vials
Leverandør: HEATHROW
HS23229B
HEATHS23229BEA 545 NOK
HEATHS23229B
Racks for cryogenic vials
Stativer for rør Kryorørstativer
Racks to store and transport cryogenic vials.
  • Embossed index numbers and squared, upper left corner aid orientation
  • Ribbed grips make handling with gloves easier
  • Made from PC, can withstand both cryogenic freezing and autoclaving
Well tabs allow one-handed opening and closing of vials that have a slot feature.
Well Ø: 12.9 mm.