Multi-function stopwatch
Leverandør: HEATHROW
HS24645
HEATHS24645EA 720 NOK
HEATHS24645
Multi-function stopwatch
Tidsur Stoppeklokker, digitale
Timing capacity: 60 minutes in ¹/₁₀₀ second increments.
  • Lightweight, dependable, accurate and easy to use
  • Large ¹/₄" display
  • Functions: split, hourly chime, calendar, alarm
Leveringsinformasjon: Supplied with neck cord and one LR44 (1,5 V) battery.