Cell spreaders, stainless steel
Leverandør: HEATHROW
HS86655
HEATHS86655EA 230 NOK
HEATHS86655
Cell spreaders, stainless steel
Utstryksspatler Celleutstryksspatler
Stainless steel spreader for use with 90 mm plates. Smooth triangular end makes spreading easy - the upturned end will not cut into agar.