Deioniser, mixed-bed, Aquademat
Leverandør: GLASWARENFABRIK KARL HECHT
40075010
HECH40075010EA 10350 NOK
HECH40075010
Deioniser, mixed-bed, Aquademat
Vannrensesystemer Ionebyttere
Ion exchanger for purified water.
  • Blue lightproof cartridge avoids growth of algae
Sertifiseringer: Conforms with DAB, EuAB.
Leveringsinformasjon: Supplied with conductivity meter with inlet hose, outlet hose and wall holder (hose connections R19 mm).