Blood sedimentation apparatus, stainless steel, MACRO, Assistent
Leverandør: GLASWARENFABRIK KARL HECHT
40367005
HECH40367005EA 3050 NOK
HECH40367005
Blood sedimentation apparatus, stainless steel, MACRO, Assistent
Pipetter Hemometerpipetter
With tilting device for rapid sedimentation.
Complete with blood sedimentation pipettes, protecting caps for pipettes, blood mixing tubes with rubber stoppers and special rubber caps.