Blood sedimentation pipettes, acc. to Westergren-Katz
Leverandør: GLASWARENFABRIK KARL HECHT
40380010
HECH40380010EA 400 NOK
HECH40380010
Blood sedimentation pipettes, acc. to Westergren-Katz
Pipetter Hemometerpipetter
Blood sedimentation pipettes for the macro method.
  • Clear glass
  • Fused blue graduation 'ASSISTENT-PERMANENT'
  • Division 0 to 200 mm: 1 mm
Sertifiseringer: DIN EN ISO 13079