Counting chamber, Bürker-Türk
Leverandør: GLASWARENFABRIK KARL HECHT
40444702
HECH40444702EA 1100 NOK
HECH40444702
Counting chamber, Bürker-Türk
Objektglass Tellekammer
Double ruling chambers for blood tests and urine analysis
  • Groups of squares in the corners divided by double lines
  • Overall size and the central square correspond to the Neubauer ruling