Enzyme test pipette, conformity certified, Assistent
Leverandør: GLASWARENFABRIK KARL HECHT
40545724
HECH40545724EA 2425 NOK
HECH40545724
Enzyme test pipette, conformity certified, Assistent
Pipetter Hemometerpipetter
Clear glass, with elongated strong tip, precision type, for partial delivery (not calibrated to tip).
  • Calibrated to deliver (Ex), short delivery time
  • Zero point at top, main points
  • Fused amber graduation 'ASSISTENT-PERMANENT'
Available with individual certificate on request (extra charge).