Assistent® Stirring Spatula
Leverandør: GLASWARENFABRIK KARL HECHT
40734010
HECH40734010EA 200 NOK
HECH40734010
Assistent® Stirring Spatula
Spatler
Designed for several uses, such as mixing samples and solutions in test tubes, resolution of substrate, additions of small reagent volumes, or releasing blood clots from the side of test tube walls.
  • Stirring rod for a wide variety of laboratory applications
Made from polystyrene, crystal clear.