Centrifuge tubes with screw cap, round bottom, AR-Glas®, Assistent®
Leverandør: GLASWARENFABRIK KARL HECHT
40976010
HECH40976010EA 2450 NOK
HECH40976010
Centrifuge tubes with screw cap, round bottom, AR-Glas®, Assistent®
Rør Sentrifugerør
Cylindrical, ungraduated, with thread GL 14.
  • Round bottom
The screw caps are gas-sterilisable up to +121 °C using ethylene oxide.