Soxhlet extractors
Leverandør: GLASWARENFABRIK KARL HECHT
41155018 41155036
HECH41155018EA 2000 NOK
HECH41155018 HECH41155036
Soxhlet extractors
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Borosilicate glass.
  • Excellent temperature resistance
  • Excellent temperature resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.