Staining trough
Leverandør: GLASWARENFABRIK KARL HECHT
41201042
HECH41201042EA 3725 NOK
HECH41201042
Staining trough
Krukker Fargetrau
Staining system contains staining jar with ground-in cover with knob made of glass, rack and handle made from stainless steel.
Suitable for 26 mm wide microscopic slides. Ideal for serial stainings, especially for the method according to Papanicolaou.