Draining rack
Leverandør: GLASWARENFABRIK KARL HECHT
41238010
HECH41238010EA 1125 NOK
HECH41238010
Draining rack
Stativer for rør Tørkestativer
Drying rack made of Plexiglas®, for 12 microscope slides.
The microscope slides are placed vertically on replaceable blotting paper. Two packs of paper can be stored in the base of the drying rack.
Leveringsinformasjon: Supplied with two packs of paper.