Microscope slide rack
Leverandør: GLASWARENFABRIK KARL HECHT
41240010
HECH41240010EA 430 NOK
HECH41240010
Microscope slide rack
Stativer for rør Fargestativer
Racks, made of Plexiglas®, for microscopic slides, with numbering 1 to 12 and field for inscriptions.