Bunsen burner
Leverandør: GLASWARENFABRIK KARL HECHT
41385010
HECH41385010EA 1925 NOK
HECH41385010
Bunsen burner
Termostater Gassbrennere
All gas bunsen burner with air regulation, needle valve and adjustable nozzle.
  • Height of approx. 160 mm
  • Tube Ø of approx. 13 mm