Gas burners, Teclu
Leverandør: GLASWARENFABRIK KARL HECHT
41393001
HECH41393001EA 2025 NOK
HECH41393001
Gas burners, Teclu
Termostater Gassbrennere
Gas burners according to Teclu, with stopcock and pilot flame.
  • Tube ~13 mm Ø
  • Height ~160 mm
  • Base ~80 mm Ø