Policemen, rubber
Leverandør: GLASWARENFABRIK KARL HECHT
41596010
HECH41596010EA 37 NOK
HECH41596010
Policemen, rubber
Skrapere
Spade shaped wiper made from natural rubber.
Int. Ø: 5 mm.
Bestillingsinformasjon: Supplied without glass rod.