Flasks acc. to Kjeldahl, with beaded rim
Leverandør: GLASWARENFABRIK KARL HECHT
41870024
HECH41870024EA 1075 NOK
HECH41870024
Flasks acc. to Kjeldahl, with beaded rim
Kolber Kjeldahlkolber
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.
Sertifiseringer: DIN 12360