Erlenmeyer flasks, narrow or wide neck, with beaded rim
Leverandør: GLASWARENFABRIK KARL HECHT
41940060
HECH41940060EA 1075 NOK
HECH41940060
Erlenmeyer flasks, narrow or wide neck, with beaded rim
Kolber Erlenmeyerkolber
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.