Digital timer

Leverandør: GLASWARENFABRIK KARL HECHT
41977060
HECH41977060EA 605 NOK
HECH41977060
Digital timer
Tidsur Tidsur, digital
Digital interval timer with large display.

  • LCD display, 4 digits showing minutes and seconds
  • Countdown 99 min 59 s with acoustic signal
  • Countdown in seconds

Weight: Approx. 70 g incl. battery

Leveringsinformasjon: Supplied with magnet, stand and battery (AAA 1,5 V).
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR