Volumetric flasks, class B, white markings
Leverandør: GLASWARENFABRIK KARL HECHT
42089092
HECH42089092EA 2075 NOK
HECH42089092
Volumetric flasks, class B, white markings
Kolber Målekolber
Borosilicate glass 3.3.
  • Higher accuracy than standard class B, Tolerance: 1,5 times of the official tolerance of class A
  • Calibrated to contain (marked 'In')
*Wide neck
Sertifiseringer: In accordance with DIN EN ISO 1042