Volumetric flasks, class A, trapezoidal shape, blue markings
Leverandør: GLASWARENFABRIK KARL HECHT
42110024
HECH42110024EA 410 NOK
HECH42110024
Volumetric flasks, class A, trapezoidal shape, blue markings
Kolber Målekolber
DURAN® Borosilicate glass 3.3 with PP stopper.
  • Calibrated to contain (TC, In)
  • Blue markings
  • Conformity certified (DE-M marking)
The base is considerably larger than of standard volumetric flasks with same volume, thus improved stability.
In accordance with DIN EN ISO 1042