Measuring beakers, with handle
Leverandør: GLASWARENFABRIK KARL HECHT
42281066
HECH42281066EA 1075 NOK
HECH42281066
Measuring beakers, with handle
Beger
Glass, with spout and graduation.
  • Content: about 1 liters
  • Outer Ø: approx. 90 mm
Ideal for general laboratory use.