Cavity slides for immunofluorescence
Leverandør: GLASWARENFABRIK KARL HECHT
42424010 42423010 42420010 42422010 42421010
631-9485EA 1925 NOK
631-9485 HECH42423010 HECH42420010 HECH42422010 HECH42421010
Cavity slides for immunofluorescence
Objektglass Cavity Slides
Black (or blue) epoxy-coated slides.
Specifically designed for immunofluorescence.