Test tubes,with rim
Leverandør: GLASWARENFABRIK KARL HECHT
42771015
HECH42771015EA 445 NOK
HECH42771015
Test tubes,with rim
Rør Reagensrør
FIOLAX®, neutral glass.
  • Very good chemical resistance
  • Relatively resistant against rapid temperature changes and local heating
Thin walled test tubes for a wide variety of applications.