Separating funnels, pear shaped
Leverandør: GLASWARENFABRIK KARL HECHT
42970066
HECH42970066EA 1975 NOK
HECH42970066
Separating funnels, pear shaped
Trakter Skilletrakter
Separating funnels, made of borosilicate glass, pear shaped, stopcock with solid glass plug.
  • Without graduations
  • Excellent chemical resistance
  • Excellent temperature resistance