Weather station
Leverandør: GLASWARENFABRIK KARL HECHT
43655010
HECH43655010EA 1525 NOK
HECH43655010
Weather station
Værstasjoner
This wireless weather station with sensor unit combines the features of a barometer (tendency), thermometer and hygrometer. The sensor unit can be placed at a distance of max. 100 m from the main station.
  • Radio controlled clock
  • With LCD display and LED indicator
  • Low battery warning
  • Stand-up device, wall mounting is possible
  • Range dependent on building structure and sensor unit placement
W×D×H (Station): approx. 102×33×180 mm / 230 g
W×D×H (Sensor unit): approx. 43×26×160 mm / 75 g
Leveringsinformasjon: Supplied with 3 batteries AA 1,5 V (LR 6).