Urine collection container
Leverandør: GLASWARENFABRIK KARL HECHT
44116010
HECH44116010EA 435 NOK
HECH44116010
Urine collection container
Diagnostiske systemer Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, transparent, with white cover made of polycarbonate.
  • Good chemical resistance
  • Good temperature resiatnce
Urine collecting container, PP, with handle and embossed scale.