Imhoff Cone without Stopcock
Leverandør: GLASWARENFABRIK KARL HECHT
44275010
HECH44275010EA 3750 NOK
HECH44275010
Imhoff Cone without Stopcock
Trakter Immhoff-trakter
Made from borosilicate 3.3 glass.
  • Very good chemical resistance
  • Excellent thermal resistance
  • With frosted label and scale
  • Fused white graduation
The sedimentation cone with graduation according to Imhoff is used for the quantification of sediments from a suspension.
Sertifiseringer: Produced according to DIN 12672.