Spray bottles
Leverandør: GLASWARENFABRIK KARL HECHT
49119010
HECH49119010EA 255 NOK
HECH49119010
Spray bottles
Flasker Sprayflasker
PE spray bottles with adjustable jet.
  • Translucent
  • Good chemical resistance
  • Good temperature resistance