Hei-CHILL 3000 Circulating Chillers
Leverandør: Heidolph Instruments GmbH & Co.KG
591-01700-00
HEID591-01700-00EA 0 NOK
HEID591-01700-00
Hei-CHILL 3000 Circulating Chillers
Termostatbad Termostatbad, lukket Termostatbad med kjøling
Extensive range of services for challenging temperature control tasks with a cooling capacity of 3000 W.
  • Adjustable bypass for pressure limiting
  • Fill opening at the top, drain tap at the rear
  • Electronic level indicator and below level alarm
  • SmartCool system for energy-saving digital cold control including automatic compressor function
  • Clear, manageable controls, large TFT display and membrane keyboard
  • USB interface and alarm contact as standard features
Integrated program controller with 50 segments, can be divided between 5 programs.