Peltier Circulator, Hei-CHILL
Leverandør: Heidolph Instruments GmbH & Co.KG
591-01910-00
HEID591-01910-00EA 68325 NOK
HEID591-01910-00
Peltier Circulator, Hei-CHILL
Termostatbad Termostatbad, lukket Termostatbad med kjøling
With a high cooling capacity of 600 W and a heating power being almost twice as high, the Hei-CHILL Peltier thermostat is suitable for a large number of temperature control tasks.
  • 6 L tank volume for temperature stability, even under sustained load
  • Precise temperature regulation and stability of ≤0,05 °C
  • Operating temperature range of −10 to +70 °C
  • Modern 2.83 in. touch display
  • Ethernet, RS 232 and RS 485 interfaces for the integration in processing solutions
The thermostat can be integrated in processes by means of the Hei-PROCESS solutions usingMore Product Information