Rack for sedimentation plates
Leverandør: MERCK LIFE SCIENCE
1465190001
HEIP1465190001EA 2675 NOK
HEIP1465190001
Rack for sedimentation plates
Stativer for rør Petriskåldispenser
High-quality stainless steel sedimentation plate rack suitable for isolators and cleanroom applications (resistant against H₂O₂ and disinfectants).
  • Locking device aids safe, convenient handling
  • Autoclavable