FEP Replacement Tube
Leverandør: HELLMA
040-101-722
HELL040-101-722EA 720 NOK
HELL040-101-722
FEP Replacement Tube
Kyvetter Spektrofotometerkyvetter
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
  • FEP replacement tube: 500 mm long, with one long screw connection
  • Outer Ø: 1,9 mm
  • Inner Ø: 1,1 mm
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.
Chemically resistant and thermally bonded: Hellma® precision cells are thermally bonded and not glued, which gives them a very high resistance to