TrayCell® Spectrophotometer Cuvettes
Leverandør: HELLMA
105810-A3-V1-4
HELL105810-A3-V1-4EA 0 NOK
HELL105810-A3-V1-4
TrayCell® Spectrophotometer Cuvettes
Kyvetter Spektrofotometerkyvetter
The TrayCell is a fiber-optic ultra-micro measuring cell designed for the UV/Vis analysis of DNA/RNA and proteins. The
dimensions of the TrayCell are equivalent to a standard cuvette in order to work in most spectrophotometers.
  • Suitable for almost any current spectrophotometers
  • Ideally suited for very small
  • During measurements, the TrayCell shows excellent reproducibility
Trouble-free measurement of the sample at different optical path lengths simply by exchanging the cap (caps with dilution factors: 5, 10, 50 and 100).
Fast and simple cleaning ofMore Product Information