Cuvette rack
Leverandør: HELLMA
3001.10
HELL3001.10EA 470 NOK
HELL3001.10
Cuvette rack
Stativer for rør Kyvettestativer
Polypropylene.
Rack for 16 square cells with 10 mm path length with inclined profile to aid cell handling.