Cleaning Holder for Cells, Hellma Analytics
Leverandør: HELLMA
325.000
HELL325.000EA 2075 NOK
HELL325.000
Cleaning Holder for Cells, Hellma Analytics
Kyvetter Spektrofotometerkyvetter
Hellma® Cleanassist, plastic cell holder for 4 cells with 10 mm path length.
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers. Highest precision: Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.
Chemically resistant and thermally bonded: Hellma® precision cells are thermally bonded and not glued, which gives them a very high resistance to temperatures and chemicals. This prevents unintentional leakage of