Magnetic Stir Bar, PTFE-Coated
Leverandør: HELLMA
332-300-VE10
HELL332-300-VE10EA 1175 NOK
HELL332-300-VE10
Magnetic Stir Bar, PTFE-Coated
Omrørere Magnetrørerstaver
For use with cell types: 109.000, 109.000F, 109.004, 109.004F, 119.000, 119.000F, 119.004, and 119.004F.
  • PTFE-coated
  • Length: 6 to 7 mm
  • 3 to 4 mm Ø