Mikroplater, kvarts
Leverandør: HELLMA
730009-44
HELL730009-44EA 77875 NOK
HELL730009-44
Mikroplater, kvarts
Mikroplater Mikroplater, kvarts
Microplates are increasingly being used in areas such as clinical chemistry and pharmaceutical research. Plastic microplates however are not suitable for all purposes. If chemical resistance, UV transmission, microscopic observation or thermal stability are required, it is better to use quartz microplates. These can be provided to the same basic dimensions as the plastic plates.
The upper part of the microplate is attached directly to the synthetic quartz base using the process of direct fusion. The result of this joining process, which does not use glue or sinter glass is an evenness ofMore Product Information