Replacement Air Filter for ULT Freezers
Leverandør: Thermo Scientific
332297H02
HERA332297H02EA 2125 NOK
HERA332297H02
Replacement Air Filter for ULT Freezers
Frysere Frysere, ultra-
Replacement air filter for ultra-low temperature chest freezers.
Routine maintenance is critical for proper ultra-low temperature (ULT) freezer operation. Replacing your ULT's old air filter enables the necessary airflow to the unit's refrigeration system.